Order Entry
Canada
ContactUsLinkComponent
SYK (phospho Tyr323) Cell Based ELISA Kit
SYK (phospho Tyr323) Cell Based ELISA Kit
Catalog # 77207-054
Supplier:  Avantor
CAS Number:  
undefined
SYK (phospho Tyr323) Cell Based ELISA Kit
Catalog # 77207-054
Supplier:  Avantor
CAS Number:  

Specifications

  • Detection method:
    Fluorometric
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    >5000 Cells
  • Storage temperature:
    4 °C
  • Regulatory status:
    RUO
  • Cat. no.:
    77207-054

Specifications

About this item

This SYK (phospho Tyr323) Cell Based ELISA kit allows for the detection of SYK (phospho Tyr323) and the effects that certain stimulation conditions have on SYK (phospho Tyr323) expression in different cell lines. Qualitative determination of SYK (phospho Tyr323) concentration is achieved by an indirect ELISA format. In essence, SYK (phospho Tyr323) is captured by Anti-SYK (phospho Tyr323) Antibodies while Dye 1-conjugated and Dye 2-conjugated secondary antibodies bind the Fc region of the primary antibody. Through this binding, the dye conjugated to the secondary antibody can emit light at a certain wavelength given proper excitation, hence allowing for a fluorometric detection method. A monoclonal antibody specific for human GAPDH is included to serve as an internal positive control in normalizing the target RFU values. Anti-SYK antibody is also included for normalization purposes. The RFU values obtained for the non-phosphorylated target can be used to normalize the RFU value for the phosphorylated target.