Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Human STAT6 ELISA Kit

Catalog # 77203-346
Supplier:   Antibodies.com
 :  A310566-96
Human STAT6 ELISA Kit
Catalog # 77203-346
Supplier:  Antibodies.com
CAS Number:  
Human STAT6 ELISA Kit
Human STAT6 ELISA Kit
CAS Number:  
Price
$934.05
Log in for availability
Each
 
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Human STAT6 ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human STAT6 in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Detection range: 100 to 1600 ng/L
  • Sensitivity: 7.98 ng/L
  • Sample type: Serum, plasma or other biological fluids
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%

Human STAT6 ELISA kit (A310566) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human STAT6 in An antibody specific for STAT6 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the STAT6 present in each sample is bound to the wells by the immobilized antibody. Biotinylated Anti-STAT6 antibody, which also binds the STAT6 present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-STAT6 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-STAT6 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Color develops in proportion to the amount of STAT6 captured in each well. The color development is stopped by addition of stop solution which changes the color from blue to yellow and the intensity of the color is then measured. The concentration of STAT6 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

 : This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, streptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.

Specifications

Specifications

  • Cat. No.:
    77203-346
  • Detection method:
    Colorimetric
  • Sensitivity:
    7.98 ng/L
  • Sample type:
    Serum, plasma or other biological fluids
  • Sample volume:
    40 μl
  • Detection range:
    100 - 1600 ng/L
  • Storage temperature:
    4 °C
  • Shelf life:
    Store for 6 months at 4 °C
  • Regulatory status:
    RUO
  • Time to Results:
    1 h 30 min

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.