Order Entry
Canada
Orders LinkContactUsLinkComponent
Human SEMA4C/SEMAI ELISA Kit

Human SEMA4C/SEMAI ELISA Kit

Human SEMA4C/SEMAI ELISA kit, 96 tests
Catalog # 77173-778
Supplier:   Antibodies.com
Supplier #:  A313425-96
Human SEMA4C/SEMAI ELISA Kit
Catalog # 77173-778
Supplier:  Antibodies.com
CAS Number:  
Human SEMA4C/SEMAI ELISA Kit
Human SEMA4C/SEMAI ELISA Kit

Human SEMA4C/SEMAI ELISA kit, 96 tests

CAS Number:  
Price
$934.05
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Human SEMA4C/SEMAI ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human SEMA4C/SEMAI in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Detection range: 50 to 800 ng/L
  • Sensitivity: 3.14 ng/L
  • Sample type: Serum, plasma or other biological fluids
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%

Human SEMA4C/SEMAI ELISA kit (A313425) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human SEMA4C/SEMAI in serum, plasma or other biological fluids. An antibody specific for SEMA4C/SEMAI has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the SEMA4C/SEMAI present in each sample is bound to the wells by the immobilized antibody. Biotinylated Anti-SEMA4C/SEMAI antibody, which also binds the SEMA4C/SEMAI present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-SEMA4C/SEMAI antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-SEMA4C/SEMAI antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Color develops in proportion to the amount of SEMA4C/SEMAI captured in each well. The color development is stopped by addition of stop solution which changes the color from blue to yellow and the intensity of the color is then measured. The concentration of SEMA4C/SEMAI in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, streptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.

Specifications

Specifications

  • Description:
    Human SEMA4C/SEMAI ELISA kit
  • Size:
    96 tests
  • Primary antibody reactivity:
    Human
  • Sample type:
    Serum, plasma or other biological fluids
  • Sample volume:
    40 μl
  • Detection range:
    50 - 800 ng/L
  • Storage temperature:
    4 °C
  • Shelf life:
    Store for 6 months at 4 °C
  • Assay Type:
    Sandwich
  • Time to Results:
    1 h 30 min
  • Cat. No.:
    77173-778
  • Assay duration:
    One step
  • Detection method:
    Colorimetric
  • Sensitivity:
    3.14 ng/L
  • Format:
    Pre-coated
  • Target protein:
    SEMA4C/SEMAI
  • Regulatory status:
    RUO

Specifications