- Assay duration:Multiple steps
- Assay Type:Indirect
- Format:Pre-coated
- Primary antibody reactivity:HumanMouse
- Target protein:PHCA
- Description:PHCA cell based ELISA kit
- Size:96 tests
- Sample type:Adherent cells and suspension cells.
- Detection method:Colorimetric
- Time to Results:4 h 30 min
- Shelf life:Store for 6 months at 4 °C
- Detection range:>5000 Cells
- Storage temperature:4 °C
- Regulatory status:RUO
- Cat. no.:77214-568
This PHCA Cell Based ELISA kit allows for the detection of PHCA and the effects that certain stimulation conditions have on PHCA expression in different cell lines. Qualitative determination of PHCA concentration is achieved by an indirect ELISA format. In essence, the PHCA is captured by Anti-PHCA Antibodies which in turn are detected by HRP-conjugated secondary antibodies. Through this binding, the HRP enzyme (conjugated to the secondary antibody) can catalyze a colorimetric reaction upon substrate addition. Due to the qualitative nature of this PHCA Cell Based ELISA kit, multiple normalization methods are described: 1) Anti-GAPDH antibody is included to serve as an internal positive control in normalizing the target absorbance values. 2) Following the colorimetric measurement of HRP activity via substrate addition, the Crystal Violet whole-cell staining method is used to determine cell density. After staining, the results can be analyzed by normalizing the absorbance values to cell amounts, by which the plating difference can be adjusted.