Order Entry
Canada
ContactUsLinkComponent
Mouse Neogenin ELISA Kit
Mouse Neogenin ELISA Kit
Catalog # 77201-126
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Mouse Neogenin ELISA Kit
Catalog # 77201-126
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A312076-96
CAS Number:  

Specifications

  • Assay duration:
    One step
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    Neogenin
  • Description:
    Mouse Neogenin ELISA kit
  • Size:
    96 tests
  • Sample type:
    Serum, plasma or other biological fluids
  • Detection method:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    150 - 2400 ng/L
  • Storage temperature:
    4 °C
  • Sample volume:
    40 μl
  • Sensitivity:
    9.75 ng/L
  • Regulatory status:
    RUO
  • Cat. no.:
    77201-126

Specifications

About this item

Mouse Neogenin ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Neogenin in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Detection range: 150 to 2400 ng/L
  • Sensitivity: 9.75 ng/L
  • Sample type: Serum, plasma or other biological fluids
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%

Mouse Neogenin ELISA kit (A312076) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse Neogenin in serum, plasma or other biological fluids. An antibody specific for Neogenin has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Neogenin present in each sample is bound to the wells by the immobilized antibody. Biotinylated Anti-Neogenin antibody, which also binds the Neogenin present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-Neogenin antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-Neogenin antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Color develops in proportion to the amount of Neogenin captured in each well. The color development is stopped by addition of stop solution which changes the color from blue to yellow and the intensity of the color is then measured. The concentration of Neogenin in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, streptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.