Human BD-2 ELISA Kit
Catalog # 77204-216
Supplier: Avantor
CAS Number:
Specifications
- Sample type:Plasma, serum and lysates
- Detection method:Colorimetric
- Time to Results:4 h 30 min
- Shelf life:Store for 6 months at 4 °C
- Detection range:8 - 1000 pg/ml
- Storage temperature:4 °C
- Regulatory status:RUO
- Cat. no.:77204-216
Specifications
About this item
This Human BD-2 ELISA kit applies a technique called a quantitative sandwich immunoassay. It allows for the detection and quantification of endogenous levels of natural or recombinant Human BD-2 in plasma, serum, and lysates.