Human MMP-7 ELISA Kit
Catalog # 77209-382
Supplier: Avantor
CAS Number:
Specifications
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Mouse
- Target protein:MMP-7
- Description:Human MMP-7 ELISA kit
- Size:96 tests
- Sample type:Plasma, serum and lysates
- Detection method:Colorimetric
- Time to Results:4 h 30 min
- Shelf life:Store for 6 months at 4 °C
- Detection range:32 - 2000 pg/ml
- Storage temperature:4 °C
- Regulatory status:RUO
- Cat. no.:77209-382
Specifications
About this item
This Human MMP-7 ELISA kit applies a technique called a quantitative sandwich immunoassay. It allows for the detection and quantification of endogenous levels of natural or recombinant Human MMP-7 in plasma, serum, and lysates.