Human G-CSFR ELISA Kit
Catalog # 77213-962
Supplier: Avantor
CAS Number:
Specifications
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Human
- Target protein:G-CSFR
- Description:Human G-CSFR ELISA kit
- Size:96 tests
- Detection method:Chemiluminescent (Luminometer)
- Shelf life:Store for 6 months at 4 °C
- Detection range:8 - 500 pg/ml
- Storage temperature:4 °C
- Regulatory status:RUO
- Cat. no.:77213-962