To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Sample type:Serum, plasma or other biological fluids
- Detection method:Colorimetric
- Time to Results:1 h 30 min
- Shelf life:Store for 6 months at 4 °C
- Detection range:0.75 - 12 ng/ml
- Storage temperature:4 °C
- Sample volume:40 μl
- Sensitivity:0.024 ng/ml
- Regulatory status:RUO
- Cat. no.:77174-298
Specifications
About this item
Human FDPS/FPS ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human FDPS/FPS in serum, plasma, and other biological fluids.
- Higher throughput: Get results in just 90 minutess, with a single wash step
- Detection Range: 0.75 to 12 ng/ml
- Sensitivity: 0.024 ng/ml
- Sample Type: Serum, plasma or other biological fluids
- Assay Precision: Intra to Assay: CV <8%, Inter to Assay: CV <10%
Human FDPS/FPS ELISA kit (A313686) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human FDPS/FPS in serum, plasma or other biological fluids. An antibody specific for FDPS/FPS has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the FDPS/FPS present in each sample is bound to the wells by the immobilized antibody. Biotinylated anti-FDPS/FPS antibody, which also binds the FDPS/FPS present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-FDPS/FPS antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-FDPS/FPS antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Color develops in proportion to the amount of FDPS/FPS captured in each well. The color development is stopped by addition of stop solution which changes the color from blue to yellow and the intensity of the color is then measured. The concentration of FDPS/FPS in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.