Progeny Handheld Raman Analyzer
Catalog #: 77270-080
Supplier: Rigaku
New Product
Specifications
- Description:Spectrometer, handheld raman analyzer
- Laser output power:30 - 490 mW
- Electrical:14.4 V Li-ion, 3100 mAH, >5 h (battery power)
- Wavelength:1064 ±1 nm
- Spectral Range:200 - 2500 cm⁻¹
- Dimensions:2.9W×3.2D×11.8"H
- Weight:1.5 kg (3.5 lbs.)
Specifications
About this item
The Rigaku Progeny handheld 1064 nm Raman analyzer is used for raw material identification and verification.
- Accurately analyze through translucent polymer and glass materials
- Measure a wider range of materials with fluorescence-free analysis
- Flexible data transfer capabilities via WiFi, USB, or docking station
- Spectral resolution 8 - 11 cm⁻¹
Perform on-the-spot, lab-quality analysis of the widest range of solids, powders, liquids, and other substances to ensure consumer safety.
Meet regulatory compliance requirements, while streamlining workflow procedures, decrease costs, increase product quality, and bring products to market faster.
Certifications: IP 68, FDA 1040, 21 CFR, CE, ISO 9001:2015 Certified Manufacturing facility.
Ordering information: 24-month warranty.
Delivery: Supplied with an adjustable nosecone adapter, vial adapter, benzonitrile calibration reference standard, 2 Li-ion rechargeable batteries with charger, and user manual.
Packaging: Packaged in a ruggedized carry case.
Customers Who Bought This Also Bought