Order Entry
Canada
Orders LinkContactUsLinkComponent
Diluent Markers

Diluent Markers

Diluent Markers
Certificates

14 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • CA76645-490
  • CA76645-008
  • CA76645-090
  • CA76645-272
  • CA76645-332
  • CA76645-334
  • CA76645-358
  • CA76645-436
  • CA76645-196
  • CA76645-326
  • CA76645-022
  • CA76645-040
  • CA76645-166
  • CA76645-158
  • Storage temperature:
  • Store at –20 °C

Specifications

Product Family Options

Product Information

  • Volume
    Details
    Pack type
    Availability
    Price
  • CA76645-490
    FP US NGS diluent marker, 1 bp lower marker, 6000 bp upper marker
    FP US NGS diluent marker, 1 bp lower marker, 6000 bp upper marker
    25 µl
    For use with the Femto pulse system and the FP-1101 kit
    1 vial
    Log in for availability
    Log in for availability
    Each
    Each - $616.00
    $616.00
    Each
       $616.00
    Minimum order quantity is 1

    Supplier Number: FP-8101-U025
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    CA76645-490
    Storage temperature
  • CA76645-008
    NGS diluent marker (1 - 6000 bp), 1 bp lower marker; 6000 bp upper marker, store at –20 ℃
    NGS diluent marker (1 - 6000 bp), 1 bp lower marker; 6000 bp upper marker, store at –20 ℃
    2.4 ml
    For use with the fragment analyzer systems and the DNF-473 kit
    1 vial
    Log in for availability
    Log in for availability
    Each
    Each - $185.00
    $185.00
    Each
       $185.00
    Minimum order quantity is 1

    Supplier Number: DNF-374-0003
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    CA76645-008
    Storage temperature
  • CA76645-090
    Large fragment diluent marker, 1 bp lower marker, 100000 bp upper marker
    Large fragment diluent marker, 1 bp lower marker, 100000 bp upper marker
    2.4 ml
    For use with the fragment analyzer systems and the DNF-492 kit
    1 vial
    Log in for availability
    Log in for availability
    Each
    Each - $185.00
    $185.00
    Each
       $185.00
    Minimum order quantity is 1

    Supplier Number: DNF-380-0003
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    CA76645-090
    Storage temperature
  • CA76645-272
    HS small fragment diluent marker, 1 bp lower marker, 1500 bp upper marker
    HS small fragment diluent marker, 1 bp lower marker, 1500 bp upper marker
    2.4 ml
    For use with the fragment analyzer systems and the DNF-477 kit
    1 vial
    Log in for availability
    Log in for availability
    Each
    Each - $185.00
    $185.00
    Each
       $185.00
    Minimum order quantity is 1

    Supplier Number: DNF-371-0003
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    CA76645-272
    Storage temperature
  • CA76645-332
    HS NGS diluent marker (1 - 6000 bp), 1 bp lower marker; 6000 bp upper marker, store at –20 ℃
    HS NGS diluent marker (1 - 6000 bp), 1 bp lower marker; 6000 bp upper marker, store at –20 ℃
    2.4 ml
    For use with the fragment analyzer systems and the DNF-474 kit
    1 vial
    Log in for availability
    Log in for availability
    Each
    Each - $185.00
    $185.00
    Each
       $185.00
    Minimum order quantity is 1

    Supplier Number: DNF-373-0003
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    CA76645-332
    Storage temperature
  • CA76645-334
    Small fragment diluent marker, 1 bp lower marker
    Small fragment diluent marker, 1 bp lower marker
    2.4 ml
    For use with the fragment analyzer systems and the DNF-476 kit
    1 vial
    Log in for availability
    Log in for availability
    Each
    Each - $185.00
    $185.00
    Each
       $185.00
    Minimum order quantity is 1

    Supplier Number: DNF-362-0003
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    CA76645-334
    Storage temperature
    Store at –20 °C
  • CA76645-358
    HS large fragment diluent marker, 1 bp lower marker, 200000 bp upper marker
    HS large fragment diluent marker, 1 bp lower marker, 200000 bp upper marker
    2.4 ml
    For use with the fragment analyzer systems and the DNF-464 kits
    1 vial
    Log in for availability
    Log in for availability
    Each
    Each - $185.00
    $185.00
    Each
       $185.00
    Minimum order quantity is 1

    Supplier Number: DNF-381-0003
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    CA76645-358
    Storage temperature
  • CA76645-436
    HS genomic DNA diluent marker, 1 bp lower marker
    HS genomic DNA diluent marker, 1 bp lower marker
    2.4 ml
    For use with the fragment analyzer systems and the DNF-468 and DNF-488 kits
    1 vial
    Log in for availability
    Log in for availability
    Each
    Each - $185.00
    $185.00
    Each
       $185.00
    Minimum order quantity is 1

    Supplier Number: DNF-375-0003
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    CA76645-436
    Storage temperature
  • CA76645-196
    FP gDNA diluent marker, 1 bp lower marker
    FP gDNA diluent marker, 1 bp lower marker
    3 ml
    For use with the femto pulse system and the FP-1002, FP-1003, and FP-1004 kits
    1 vial
    Log in for availability
    Log in for availability
    Each
    Each - $309.00
    $309.00
    Each
       $309.00
    Minimum order quantity is 1

    Supplier Number: FP-8001-0003
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    CA76645-196
    Storage temperature
  • CA76645-326
    FP US RNA diluent marker, 15 nt lower marker
    FP US RNA diluent marker, 15 nt lower marker
    3 ml
    For use with the Femto pulse system and the FP-1201 kit
    1 vial
    Log in for availability
    Log in for availability
    Each
    Each - $309.00
    $309.00
    Each
       $309.00
    Minimum order quantity is 1

    Supplier Number: FP-8201-0003
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    CA76645-326
    Storage temperature
  • CA76645-022
    Small RNA diluent marker, 1 nt lower marker
    Small RNA diluent marker, 1 nt lower marker
    4 ml
    For use with the fragment analyzer systems and the DNF-470 kit
    1 vial
    Log in for availability
    Log in for availability
    Each
    Each - $462.00
    $462.00
    Each
       $462.00
    Minimum order quantity is 1

    Supplier Number: DNF-368-0004
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    CA76645-022
    Storage temperature
  • CA76645-040
    HS RNA diluent marker, 15 nt lower marker
    HS RNA diluent marker, 15 nt lower marker
    4 ml
    For use with the fragment analyzer systems and the DNF-472 kit
    1 vial
    Log in for availability
    Log in for availability
    Each
    Each - $309.00
    $309.00
    Each
       $309.00
    Minimum order quantity is 1

    Supplier Number: DNF-370-0004
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    CA76645-040
    Storage temperature
  • CA76645-166
    RNA diluent marker, 15 nt lower marker
    RNA diluent marker, 15 nt lower marker
    4 ml
    For use with the fragment analyzer systems and the DNF-471 kit
    1 vial
    Log in for availability
    Log in for availability
    Each
    Each - $309.00
    $309.00
    Each
       $309.00
    Minimum order quantity is 1

    Supplier Number: DNF-369-0004
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    CA76645-166
    Storage temperature
  • CA76645-158
    HS genomic DNA diluent marker, 1 bp lower marker
    HS genomic DNA diluent marker, 1 bp lower marker
    120 ml
    For use with the fragment analyzer systems and the DNF-467 kit
    1 Vial
    Log in for availability
    Log in for availability
    Each
    Each - $925.00
    $925.00
    Each
       $925.00
    Minimum order quantity is 1

    Supplier Number: DNF-375-0120
    Specifications:
    More
    Specifications
    Cat. No.
    CA76645-158
    Storage temperature
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.