ContactUsLinkComponent
Guardian™ 2000 Analog Hot Plates and Stirrers
Guardian™ 2000 Analog Hot Plates and Stirrers
Supplier:  Ohaus
Certificates
undefined
Guardian™ 2000 Analog Hot Plates and Stirrers
Supplier:  Ohaus
Specifications
  • Catalog No:
  • 77270-224
  • 77270-238
  • 77270-236
  • 77270-234
  • 77270-228
  • 77270-232
  • 77270-230
  • W×D×H:
  • 318×223×112 mm
  • 264×163×101 mm
  • 318×223×12mm
  • 264×163×101 mm
  • 264×163×101 mm
  • 264×163×101 mm
  • 318×223×12mm
  • Weight:
  • 3.9 kg