Order Entry
Canada
ContactUsLinkComponent
ZymoScript RT PreMix Kit
ZymoScript RT PreMix Kit
Catalog # 77020-264
Supplier:  Zymo Research
CAS Number:  
ZymoScript RT PreMix Kit
Catalog # 77020-264
Supplier:  Zymo Research
Supplier Number:  R3012
CAS Number:  

Specifications

  • Prep size:
    Scalable
  • Refine target molecule:
    Total RNA
  • Sample type:
    RNA preps
  • Target molecule:
    RNA
  • Description:
    ZymoScript RT PreMix kit
  • Cat. no.:
    77020-264

Specifications

About this item

ZymoScript RT PreMix is an optimized reverse transcription kit containing all the necessary components for quick and efficient cDNA synthesis.

  • Efficient and quick cDNA synthesis (in as little as 10 minutes)

ZymoScript RT PreMix is an optimized reverse transcription kit containing all the necessary components for quick and efficient cDNA synthesis. This reverse transcription mix includes random hexamer and oligo dT primers, dNTPs, RNase inhibitors, reverse transcriptase, and an optimized reaction buffer which includes a visual tracking dye for accurate reaction set-up. This is the kit of choice for analysis of RNA via 2-step RT-qPCR.