Order Entry
Canada
ContactUsLinkComponent
EZ DNA Methylation-Lightning Automation Kit
EZ DNA Methylation-Lightning Automation Kit
Catalog # 77020-292
Supplier:  Zymo Research
CAS Number:  
EZ DNA Methylation-Lightning Automation Kit
Catalog # 77020-292
Supplier:  Zymo Research
Supplier Number:  D5049
CAS Number:  

Specifications

  • Method / Format:
    Multiwell plate
  • Prep size:
    Scalable
  • Refine target molecule:
    Methylated DNA
  • Sample type:
    DNA preps
  • Target molecule:
    DNA
  • Description:
    EZ DNA methylation-lightning automation kit
  • Size:
    96 preps
  • Cat. no.:
    77020-292

Specifications

About this item

Automation-specific streamlined design for high-throughput bisulfite conversion of DNA for methylation analysis.

  • Ready to Use: Preassembled buffers allow for lightning-fast library preparation in as little as 4 hours without compromising quality

The EZ DNA Methylation-Lightning Automation Kit is a high-throughput DNA bisulfite conversion kit designed for streamlining automation workflows. It is recommended for several downstream Illumina® Infinium® products such as MethylationEPIC, HTS iSelect® Methyl Custom, and Mouse Methylation BeadChip Arrays. This automation-specific kit features optimized buffer volumes for consistent performance on all open platform liquid handlers and magnetic bead transfer automated systems.