ZymoPURE™ Wash Buffer 1
Catalogue # 77020-328
Fournisseur: Zymo Research
Numéro CAS:
Spécifications
- Method / Format:Multiwell plate
- Prep size:Giga
- Refine target molecule:Plasmid (endotoxin-free)
- Sample type:Bacteria
- Target molecule:Plasmid
- Description:ZymoPURE™ wash buffer 1
- Size:500 ml
- Cat. no.:77020-328
Spécifications
À propos de cet article
Wash buffer designed to be used with our ZymoPURE™ Plasmid Purification Kits.
- Efficiently isolate transfection-grade plasmid DNA from E.coli
ZymoPURE™ Wash 1 is designed to be used with our ZymoPURE™ Plasmid Purification Kits, which is the fastest and simplest method available to efficiently isolate transfection-grade plasmid DNA from E.coli.