To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Method / Format:Magnetic beads
- Prep size:Scalable
- Refine target molecule:Amplified/modified DNA
- Sample type:PCR and other enzymatic reactions
- Target molecule:DNA
- Description:DNA clean and concentrator MagBead kit
- Cat. no.:77020-260
Specifications
About this item
Magnetic bead based clean-up kit that can purify DNA from PCR, enzymatic reactions, impure extractions, and other sources.
- Rapid and scalable protocol for low to high-throughput purification of DNA from PCR, enzymatic reactions, library preparations, and other sources
The DNA clean and concentrator MagBead kit is magnetic bead based clean-up kit that can purify DNA from PCR, enzymatic reactions, impure extractions, and other sources. The MagBinding bead technology provides a rapid and scalable workflow that can be adapted for low to high-throughput automated method for purification and concentration of DNA.