To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Method / Format:Column (silica-based)
- Prep size:Mini
- Refine target molecule:Plasmid (endotoxin-free)
- Sample type:Bacteria
- Target molecule:Plasmid
- Description:Zymo-Spin II-PX columns
- Size:50 pack
- Binding capacity:100 µg
- Cat. no.:77020-324
The versatile Zymo-Spin II-PX column can be used in microcentrifuges or on vacuum manifolds for the purification of plasmid DNA.
- Allows for purification of up to 100 µg of plasmid DNA in ≥25 µl eluate when used in combination with ZymoPURE™ plasmid prep buffers
Available as a refill for the ZymoPURE™ plasmid miniprep kit. The versatile Zymo-Spin II-PX column can be used in microcentrifuges or on vacuum manifolds for the purification of plasmid DNA. The Zymo-Spin II-PX features durable polypropylene construction and a unique silica-based matrix. Capacity is 800 µl.