Zymo-Spin VP-X Column
Catalog # 77020-278
Supplier: Zymo Research
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Method / Format:Column (silica-based)
- Prep size:Maxi
- Refine target molecule:Plasmid (endotoxin-free)
- Sample type:Bacteria
- Target molecule:Plasmid
- Description:Zymo-Spin VP-X column assembly with 15 and 50 ml reservoir
- Size:5 pack
- Binding capacity:3.0 mg
- Cat. no.:77020-278
Specifications
About this item
The versatile Zymo-Spin V-PX with 15 and 50 ml reservoirs can be used in centrifuges or on vacuum manifolds for the purification of plasmid DNA.
- Allows for purification of up to 3 mg of plasmid DNA in ≥300 µl eluate when used in combination with ZymoPURE™ plasmid prep buffers
Available as a refill for the ZymoPURE™ II plasmid maxiprep kit. The Zymo-Spin V-PX features durable polypropylene construction and a unique silica-based matrix. Capacity with reservoir assembly is 65 ml.