Anti-PLCXD2 Rabbit Polyclonal Antibody
Catalog # 76762-090
Supplier: Prosci
Specifications
- Antibody type:Primary
- Antigen name:PI-PLC X domain-containing protein 2
- Antigen symbol:PLCXD2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:257068
- Antigen synonyms:PI-PLC X domain-containing protein 2|PLCXD2
- Storage buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-305 of human PLCXD2 (NP_001172035.1).
- Purification:Affinity purification
- Cat. no.:76762-090
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: PLCXD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat