Anti-PPP3R2 Rabbit Polyclonal Antibody
76752-218
: Prosci
- Antibody type:Primary
- Antigen name:Protein phosphatase 3 regulatory subunit B, beta
- Antigen symbol:PPP3R2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:5535
- Antigen synonyms:beta isoform|regulatory subunit B|19kDa|type II)|protein phosphatase 3 (formerly 2B)|calcineurin B-like protein|PPP3RL|calcineurin BII|calcineurin subunit B type 2|calcineurin B|regulatory subunit B (19kD)|type II (19kDa)|protein phosphatase 3 regulatory subunit B beta isoform|CBLP|beta isoform (calcineurin B|protein phosphatase 2B regulatory subunit 2|CNBII
- Storage buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-173 of human PPP3R2 (NP_671709.1).
- Purification:Affinity purification
- Cat. no.:76752-218
- Packaging:Vial
Type: Primary
Antigen: PPP3R2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat