Anti-NDUFAF5 Rabbit Polyclonal Antibody
Catalog # 76751-344
Supplier: Prosci
Specifications
- Antibody type:Primary
- Antigen name:NADH:ubiquinone oxidoreductase complex assembly factor 5
- Antigen symbol:NDUFAF5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:79133
- Antigen synonyms:NADH dehydrogenase (ubiquinone) complex I|arginine-hydroxylase NDUFAF5|bA526K24.2|NADH dehydrogenase [ubiquinone] 1 alpha subcomplex assembly factor 5|dJ842G6.1|probable methyltransferase C20orf7|mitochondrial|putative methyltransferase NDUFAF5|MC1DN16|C20orf7|assembly factor 5
- Storage buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 196-345 of human NDUFAF5 (NP_077025.2).
- Purification:Affinity purification
- Cat. no.:76751-344
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: NDUFAF5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat