Anti-AP2A1 Rabbit Polyclonal Antibody
Catalog # 76754-440
Supplier: Prosci
Specifications
- Antibody type:Primary
- Antigen name:Adaptor-related protein complex 2, alpha 1 subunit
- Antigen symbol:AP2A1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:160
- Antigen synonyms:adaptor protein complex AP-2 subunit|AP-2 complex subunit alpha-1|adapter-related protein complex 2 alpha-1 subunit|CLAPA1|AP2A1|alpha A|AP2-ALPHA|adaptin|alpha 1 subunit|ADTAA|adaptor-related protein complex 2|100 kDa coated vesicle protein A
- Storage buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human AP2A1 (NP_055018.2).
- Purification:Affinity purification
- Cat. no.:76754-440
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: AP2A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat