Anti-ADAD1 Rabbit Polyclonal Antibody
76761-974
: Prosci
- Antibody type:Primary
- Antigen name:Adenosine deaminase domain containing 1
- Antigen symbol:ADAD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:132612
- Antigen synonyms:testis tissue sperm-binding protein Li 92mP|testis nuclear RNA-binding protein|adenosine deaminase domain-containing protein 1|Tenr|adenosine deaminase domain containing 1 (testis-specific)
- Storage buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human ADAD1 (NP_001152757.1).
- Purification:Affinity purification
- Cat. no.:76761-974
- Packaging:Vial
Type: Primary
Antigen: ADAD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat