Anti-PNPLA2 Rabbit Polyclonal Antibody
Catalog # 76756-984
Supplier: Prosci
Specifications
- Antibody type:Primary
- Antigen name:Patatin-like phospholipase domain containing 2
- Antigen symbol:PNPLA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:57104
- Antigen synonyms:desnutrin|TTS-2.2|pnpla2|patatin-like phospholipase domain containing 2|ATGL|transport-secretion protein|triglyceride hydrolase|1110001C14Rik|FP17548|0610039C21Rik|adipose triglyceride lipase
- Storage buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 265-504 of human PNPLA2 (NP_065109.1).
- Purification:Affinity purification
- Cat. no.:76756-984
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: PNPLA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat