Anti-SF3B3 Rabbit Polyclonal Antibody
76761-892
: Prosci
- Antibody type:Primary
- Antigen name:Splicing factor 3b, subunit 3, 130kDa
- Antigen symbol:SF3B3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:23450
- Antigen synonyms:KIAA0017|SF3b130|SF3B3|SAP130|STAF130|RSE1
- Storage buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 948-1217 of human SF3B3 (NP_036558.3).
- Purification:Affinity purification
- Cat. no.:76761-892
- Packaging:Vial
Type: Primary
Antigen: SF3B3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat