Anti-OPA3 Rabbit Polyclonal Antibody
50 µl, Vial
Catalog # 76749-680
Supplier: Prosci
Supplier #: 19-518
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Type: Primary
Antigen: OPA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Size:50 µl
- Packaging:Vial
- Western blot:Yes
- Clonality:Polyclonal
- Purification:Affinity purification
- Antibody type:Primary
- Form:Liquid
- Isotype:IgG
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Host:Rabbit
- Concentration:Batch dependent
- Shipping temperature:20
- Antigen name:Outer mitochondrial membrane lipid metabolism regulator OPA3
- Cat. No.:76749-680
- Reactivity:Human|Rat|Mouse
- Gene ID:80207
- Antigen synonyms:optic atrophy 3 protein|MGA3|OPA3 outer mitochondrial membrane lipid metabolism regulator|with chorea and spastic paraplegia)|Optic atrophy 3 (Iraqi-Jewish 'optic atrophy plus')|optic atrophy 3 (autosomal recessive
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 25-179 of human OPA3 (NP_079412.1).
- Conjugation:Unconjugated
- Antigen symbol:OPA3
- Storage buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.



