Anti-IL1RAPL1 Rabbit Polyclonal Antibody
Catalog # 76764-364
Supplier: Prosci
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 1 receptor accessory protein like 1
- Antigen symbol:IL1RAPL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:11141
- Antigen synonyms:TIGIRR-2|three immunoglobulin domain-containing IL-1 receptor-related 2|interleukin-1 receptor accessory protein-like 1|IL1RAPL|mental retardation|MRX1MRX21|MRX34|IL1RAPL-1|OPHN4|IL-1-RAPL-1|interleukin 1 receptor-8|X-linked interleukin-1 receptor accessory protein-like 1|IL-1RAPL-1|IL1R8|X-linked 1oligophrenin-4
- Storage buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 130-210 of human IL1RAPL1 (NP_055086.1).
- Purification:Affinity purification
- Cat. no.:76764-364
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: IL1RAPL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat