SARS-CoV-2 (COVID-19) Spike Q493R G496S Q498R N501Y Y505H Peptide (Omicron)
Catalog # 77133-624
Supplier: Prosci
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Synthetic
- Species:SARS-CoV-2
- Size:0.05 mg
- Storage conditions:Store SARS-CoV-2 (COVID-19) Spike Q493R G496S Q498R N501Y Y505H Peptide (Omicron) at −20 °C, stable for one year.
- Gene ID:43740568
- Protein synonyms:Spike protein|Omicron Variant|Surface Glycoprotein|SARS-CoV-2 Omicron Variant Spike antibody: Severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2)
- Protein/peptide name:Spike
- Concentration:200 µg/ml
- Formulation:Liquid. PBS pH 7.2 (10 mM NaH₂PO₄, 10 mM Na₂HPO₄, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide.
- Shipping temperature:20
- Tested applications:Blocking
- Cat. no.:77133-624