SARS-CoV-2 (COVID-19, 2019-nCoV) Recombinant Spike Protein (from HEK293 Cells)
Catalog #: 77131-628
Supplier: Prosci
New Product
CAS Number:
Specifications
- Gene ID:43740578
- Packaging:Vial
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:SARS-CoV-2
- Conjugation:Unconjugated
- Size:0.05 mg
- Storage conditions:Store in working aliquots at −70 °C. Avoid freeze/thaw cycles. When working with proteins care should be taken to keep recombinant protein at a cool and stable temperature.
- Protein synonyms:S glycoprotein RBDSpike glycoprotein Receptor-binding domainDelta variant spike RBDSpike protein RBDS protein RBD
- Protein/peptide name:Spike
- Purity:>90% as determined by Bis-Tris PAGE
- Molecular weight:120 kDa
- Sequence:Val 16 - Arg 685 (T19R, G142D, EF156-157del, R158G, L452R, T478K, D614G, P681R)-His-Avi
- Formulation:Liquid. PBS (pH 7.4).
- Shipping temperature:Dry ice
- Tested applications:ELISA, WB, SDS-PAGE
Specifications
About this item
Customers Who Bought This Also Bought