SARS-CoV-2 Recombinant Spike RBD (R346K, E484K, N501Y, D614G, P681H) Protein (from Mammalian Cells)
Catalog #: 77130-988
Supplier: Prosci
New Product
CAS Number:
Specifications
- Packaging:Vial
- Protein/peptide type:Recombinant
- Source:Mammalian cells
- Species:SARS-CoV-2
- Conjugation:Unconjugated
- Size:0.1 mg
- Storage conditions:Use a manual defrost freezer and avoid repeated freeze thaw cycles.Store at 2...8 °C for one week .Store at −20...−80 °C for twelve months from the date of receipt.
- Protein synonyms:Colombia Mu Variant( B.1.621)
- Protein/peptide name:Spike RBD (R346K, E484K, N501Y, D614G, P681H)
- Purity:>90% as determined by SDS-PAGE
- Molecular weight:35 kDa
- Formulation:Lyophilized. Supplied as solution form in PBS, pH 7.5 or lyophilized from PBS, pH 7.5.
- Shipping temperature:Room temperature
Specifications
About this item
Customers Who Bought This Also Bought