Rhesus Monkey Recombinant OX40 Ligand/TNFSF4 Protein (from HEK293 Cells)
Catalog #: 77129-722
Supplier: Prosci
New Product
CAS Number:
Specifications
- Packaging:Vial
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Rhesus monkey
- Conjugation:Unconjugated
- Size:0.1 mg
- Storage conditions:Lyophilized Protein should be stored at −20 °C or lower for long term storage. Upon reconstitution, working aliquots should be stored at −20 or −70 °C. Avoid repeated freeze-thaw cycles.
- Protein synonyms:TNFSF4CD134LTXGP1OX40LGlycoprotein Gp34CD134 ligandCD252
- Protein/peptide name:OX40 Ligand/TNFSF4
- Purity:>90% as determined by SDS-PAGE
- Molecular weight:42.5 kDa
- Sequence:Gln 51 - Leu 183
- Formulation:Lyophilized. 50 mM Tris, 100 mM Glycine, pH 7.5.
- Shipping temperature:Room temperature
- Tested applications:ELISA, WB
Specifications
About this item
Customers Who Bought This Also Bought