Anti-POLR2F Rabbit Polyclonal Antibody
76761-026
: Prosci
- Antibody type:Primary
- Antigen name:RNA polymerase II subunit F
- Antigen symbol:POLR2F
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:5435
- Antigen synonyms:RPB14.4|and III 14.4 kDa polypeptide|RNA polymerases I|RPABC2|DNA-directed RNA polymerases I|polymerase (RNA) II subunit F|and III subunit RPABC2|II|RPABC14.4|polymerase (RNA) II (DNA directed) polypeptide F|RNA Polymerase II subunit 14.4 kD|and III subunit ABC2|RPC15|HRBP14.4|POLRF|RPB6|DNA-directed RNA polymerase II subunit F
- Modification:None
- Storage buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:Store at –20...+730 °C. Avoid freeze / thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-127 of human POLR2F (NP_068809.1).
- Purification:Affinity purification.
- Cat. no.:76761-026
- Packaging:Vial
Type: Primary
Antigen: POLR2F
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse