Anti-KANSL1 Rabbit Polyclonal Antibody
Catalog # 76748-660
Supplier: Prosci
Specifications
- Antibody type:Primary
- Antigen name:KAT8 regulatory NSL complex subunit 1
- Antigen symbol:KANSL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:284058
- Antigen synonyms:NSL complex protein NSL1|MSL1v1|MSL1 homolog 1|non-specific lethal 1 homolog|KDVS|MLL1/MLL complex subunit KANSL1|CENP-36|male-specific lethal 1 homolog|centromere protein 36|KIAA1267|KAT8 regulatory NSL complex subunit 1|NSL1|hMSL1v1
- Modification:None
- Storage buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:Store at –20...+730 °C. Avoid freeze / thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 806-1105 of human KANSL1 (NP_056258.1).
- Purification:Affinity purification.
- Cat. no.:76748-660
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: KANSL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human