Anti-GFM2 Rabbit Polyclonal Antibody
76751-934
: Prosci
- Antibody type:Primary
- Antigen name:G elongation factor mitochondrial 2
- Antigen symbol:GFM2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:84340
- Antigen synonyms:mEF-G 2|RRF2mt|hEFG2|RRF2|mitochondrial elongation factor G2|mitochondrial ribosome recycling factor 2|mitochondrial|EFG2|MRRF2|MST027|RRF|MSTP027|ribosome-releasing factor 2|EF-G2mt
- Modification:None
- Storage buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:Store at –20...+730 °C. Avoid freeze / thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 520-779 of human GFM2 (NP_115756.2).
- Purification:Affinity purification.
- Cat. no.:76751-934
- Packaging:Vial
Type: Primary
Antigen: GFM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat