Anti-TRAF2 Rabbit Polyclonal Antibody
Catalog # 76764-728
Supplier: Prosci
Specifications
- Antibody type:Primary
- Antigen name:TNF receptor-associated factor 2
- Antigen symbol:TRAF2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:7186
- Antigen synonyms:MGC:45012|TRAF2 Antibody: TRAP|TRAP3|TNF receptor-associated factor 2|E3 ubiquitin-protein ligase TRAF2
- Modification:None
- Storage buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:Store at –20...+730 °C. Avoid freeze / thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human TRAF2 (NP_066961.2).
- Purification:Affinity purification.
- Cat. no.:76764-728
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: TRAF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat