Anti-DOK4 Rabbit Polyclonal Antibody
76761-474
: Prosci
- Antibody type:Primary
- Antigen name:Docking protein 4
- Antigen symbol:DOK4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:55715
- Antigen synonyms:IRS-5|Downstream of tyrosine kinase 4|IRS5|DOK4|Docking protein 4|Insulin receptor substrate 5
- Modification:None
- Storage buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:Store at –20...+730 °C. Avoid freeze / thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 240-326 of human DOK4 (NP_060580.2).
- Purification:Affinity purification.
- Cat. no.:76761-474
- Packaging:Vial
Type: Primary
Antigen: DOK4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat