Anti-Anti Spike Glycoprotein antibody Recombinant Antibody
76749-722
: Prosci
- Antibody type:Primary
- Clonality:Recombinant
- Conjugation:Unconjugated
- ELISA:Yes
- Reactivity:Virus
- Western blot:Yes
- Size:0.1 mg
- Form:Liquid
- Antigen synonyms:Anti-2019-nCOV RBD antibody|Anti Spike glycoprotein antibody|Anti S2 antibody|Anti Spike protein S2 antibody|Anti 2019-nCOV spike antibody
- Modification:None
- Storage buffer:PBS,pH 7.5 .
- Storage temperature:Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 ...730 °C for 1-2 weeks. Store at –20…+730 °C for 12 months. Store at –80…+730 °C for long term storage.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant SARS-CoV-2 S2 protein.
- Cat. no.:76749-722
- Packaging:Vial
Type: Primary
Antigen:
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Human IgG
Reactivity: Virus