Order Entry
Canada
ContactUsLinkComponent
Anti-BMS-936558 Recombinant Antibody
Anti-BMS-936558 Recombinant Antibody
Catalog # 76762-912
Supplier:  Prosci
Anti-BMS-936558 Recombinant Antibody
Catalog # 76762-912
Supplier:  Prosci
Supplier Number:  10-495
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Clonality:
    Recombinant
  • Conjugation:
    Unconjugated
  • Isotype:
    IgG4 kappa
  • Size:
    0.1 mg
  • Antigen synonyms:
    IPH2102|BMS-986015
  • Modification:
    None
  • Storage buffer:
    PBS buffer pH 7.5 .
  • Storage temperature:
    Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 ...730 °C for 1-2 weeks. Store at –20…+730 °C for 12 months. Store at –80…+730 °C for long term storage.
  • Concentration:
    Batch dependent
  • Shipping temperature:
    20
  • Immunogen:
    Homo sapiens/KIRD2 subgroup [Homo sapiens].
  • Purification:
    >95%
  • Cat. no.:
    76762-912
  • Packaging:
    Vial

Specifications

About this item

Type: Primary
Antigen:
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG4-kappa
Reactivity: