Order Entry
Canada
Orders LinkContactUsLinkComponent
CCL3 Peptide

CCL3 Peptide

Vial, 0.1 mg
Catalog # 77133-908
Supplier:   Prosci
Supplier #:  45-379P
CCL3 Peptide
Catalog # 77133-908
Supplier:  Prosci
CAS Number:  
CCL3 Peptide

Vial, 0.1 mg

CAS Number:  
Price
$477.16
Log in for availability
Each
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Vial
  • Size:
    0.1 mg
  • Cat. No.:
    77133-908
  • Protein/peptide type:
    Synthetic
  • Protein/peptide name:
    CCL3
  • Storage conditions:
    Store at −20 °C. When reconstituted minimize freezing and thawing.
  • Gene ID:
    6348
  • Formulation:
    Lyophilized. CCL3 peptide is supplied as 100 µg of dried peptide. When the peptide is reconstituted in 200 µl water, the concentration will be 0.5 mg/ml.
  • Protein synonyms:
    G0S19-1|SCYA3|LD78ALPHA|Macrophage inflammatory protein 1-alpha|MIP-1-alpha|MIP1A|C-C motif chemokine 3
  • Conjugation:
    Unconjugated
  • Tested applications:
    Blocking
  • Shipping temperature:
    RT

Specifications