Order Entry
ContactUsLinkComponent
Human Recombinant IL-12Hp70 Protein (from HEK293 cells)
Human Recombinant IL-12Hp70 Protein (from HEK293 cells)
Catalog #: 77120-340
Supplier:  Prosci
New Product
CAS Number:  
Human Recombinant IL-12Hp70 Protein (from HEK293 cells)
Catalog #: 77120-340
Supplier:  Prosci
New Product
CAS Number:  

Specifications

  • Gene ID:
    3592
  • Packaging:
    Vial
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Conjugation:
    Unconjugated
  • Size:
    2 µg
  • Storage conditions:
    The recombinant protein is stable for at least 2 years from date of receipt at −20 °C. Reconstituted protein is stable for at least 3 months when stored in working aliquots with a carrier protein at −20 °C. As with any protein, exposing the recombinant protein to repeated freeze/thaw cycles is not recommended. When working with proteins care should be taken to keep recombinant protein at a cool and stable temperature.
  • Biological activity:
    Determined by its ability to increase IFN-γ production by anti-TCR mAb-stimulated PBMCs.
  • Protein synonyms:
    NKSF
    NK cell Stimulating Factor Chain 1
    p40: Interleukin-12 subunit beta
    Cytotoxic Lymphocyte Maturation Factor (CLMF)
    Cytotoxic Lymphocyte Maturation Factor 35 kDa subunit (CLMF p35)
    IL-12 subunit p40
    IL-12A
    IL-12 subunit p35
    IL-12B
    CTL Maturation Factor (TCMF)
    CytotoxicLymphocyte Maturation Factor 40 kDa subunit (CLMF p40)
    NK cell Stimulating Factor Chain 2
    TSF
    p35: Interleukin-12 subunit alpha
    Interleukin-12
  • Protein/peptide name:
    IL12A
  • Purity:
    ≥98% by SDS-PAGE gel and HPLC analyses
  • Sequence:
    p35 Subunit: RNLPVATPDP GMFPCLHHSQ NLLRAVSNML QKARQTLEFY PCTSEEIDHE DITKDKTSTV EACLPLELTK NESCLNSRET SFITNGSCLA SRKTSFMMAL CLSSIYEDLK MYQVEFKTMN AKLLMDPKRQ IFLDQNMLAV IDELMQALNF NSETVPQKSS LEEPDFYKTK IKLCILLHAF RIRAVTIDRV MSYLNAS

    p40 Subunit: IWELKKDVYV VELDWYPDAP GEMVVLTCDT PEEDGITWTL DQSSEVLGSG KTLTIQVKEF GDAGQYTCHK GGEVLSHSLL LLHKKEDGIW STDILKDQKE PKNKTFLRCE AKNYSGRFTC WWLTTISTDL TFSVKSSRGS SDPQGVTCGA ATLSAERVRG DNKEYEYSVE CQEDSACPAA EESLPIEVMV DAVHKLKYEN YTSSFFIRDI IKPDPPKNLQ LKPLKNSRQV EVSWEYPDTW STPHSYFSLT FCVQVQGKSK REKKDRVFTD KTSATVICRK NASISVRAQD RYYSSSWSEW ASVPCS
  • Formulation:
    Lyophilized
  • Shipping temperature:
    Room temperature

Specifications

About this item

Customers Who Bought This Also Bought