Human, Mouse Recombinant sIL-2 Receptor a Protein (from (BTI-Tn-5B1-4) High-5 Insect cells)
Catalog #: 77120-824
Supplier: Prosci
New Product
CAS Number:
Specifications
- Gene ID:3559
- Packaging:Vial
- Protein/peptide type:Recombinant
- Source:(BTI-Tn-5B1-4) Hi-5 Insect cells
- Species:HumanMouse
- Conjugation:Unconjugated
- Size:5 µg
- Storage conditions:The recombinant protein is stable for at least 2 years from date of receipt at −20 °C. Reconstituted protein is stable for at least 3 months when stored in working aliquots with a carrier protein at −20 °C. As with any protein, exposing the recombinant protein to repeated freeze/thaw cycles is not recommended. When working with proteins care should be taken to keep recombinant protein at a cool and stable temperature.
- Biological activity:Determined by its ability to increase the proliferation effect of IL-2 in murine CTLL-2 cells. In the presence of 1 ng/ml of recombinant IL-2, the expected ED50 for this effect is between 0.5 - 1.5 µg/ml.
- Protein synonyms:TAC-antigensoluble IL-2 receptorCD25 antigen
- Protein/peptide name:IL2RA
- Purity:≥98% by SDS-PAGE gel and HPLC analyses
- Sequence:ELCDDDPPEI PHATFKAMAY KEGTMLNCEC KRGFRRIKSG SLYMLCTGNS SHSSWDNQCQ CTSSATRNTT KQVTPQPEEQ KERKTTEMQS PMQPVDQASL PGHCREPPPW ENEATERIYH FVVGQMVYYQ CVQGYRALHR GPAESVCKMT HGKTRWTQPQ LICTGEMETS QFPGEEKPQA SPEGRPESET SCLVTTTDFQ IQTEMAATME TSIFTTEYQ
- Formulation:Lyophilized
- Shipping temperature:Room temperature
Specifications
About this item
Customers Who Bought This Also Bought