SARS-CoV-2 (COVID-19, 2019-nCoV) Recombinant Omicron Variant (B.1.1.529) Nucleocapsid Protein (from E. coli)
Catalog #: 77131-532
Supplier: Prosci
New Product
CAS Number:
Specifications
- Gene ID:43740575
- Packaging:Vial
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:SARS-CoV-2
- Conjugation:Unconjugated
- Size:0.1 mg
- Storage conditions:Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2...8 °C for one week . Store at −20...−80 °C for twelve months from the date of receipt.
- Protein synonyms:2019-nCoV novel coronavirus Nucleoprotein Protein2019-nCoV coronavirus NP Protein2019-nCoV np Protein
- Protein/peptide name:Nucleocapsid
- Purity:>90% as determined by SDS-PAGE
- Molecular weight:46.31 kDa
- Sequence:SARS-CoV-2 Nucleocapsid (YP_009724397.2, with mutations P13L, ERS31-33 deletion, R203K, G204R) (Met1-Ala419)
- Formulation:Lyophilized. Supplied as solution form in PBS, pH 7.5 or Lyophilized from PBS, pH 7.5
- Shipping temperature:Room temperature
Specifications
About this item
Customers Who Bought This Also Bought