SARS-CoV-2 (COVID-19) Recombinant Omicron Variant (B.1.1.529) S1 Protein (from Mammalian Cells)
Catalog # 77129-964
Supplier: Prosci
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:Mammalian cells
- Species:SARS-CoV-2
- Size:0.1 mg
- Storage conditions:Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2...8 °C for one week . Store at −20...−80 °C for twelve months from the date of receipt.
- Gene ID:43740578
- Protein synonyms:S glycoprotein RBD|Spike glycoprotein Receptor-binding domain|Delta variant spike RBD|Spike protein RBD|S protein RBD
- Protein/peptide name:Spike
- Purity:>90% as determined by SDS-PAGE
- Molecular weight:78.71 kDa
- Sequence:The target gene encoding Ser13-Arg682. Mutation points A67V, H69del, V70del, T95I, G142D, V143del, Y144del, Y145del, N211del, L212I, ins214EPE, G339D, S371L, S373P, S375F, K417N, N440K, G446S, S477N, T478K, E484A, Q493R, G496S, Q498R, N501Y, Y505H, T547K, D614G, H655Y, N679K, P681H
- Formulation:Lyophilized. Supplied as solution form in PBS, pH 7.5 or Lyophilized from PBS, pH 7.5
- Shipping temperature:RT
- Cat. no.:77129-964