SARS-CoV-2 (COVID-19) Recombinant Omicron Variant (B.1.1.529) Spike Trimer Protein (from Mammalian Cells)
Catalog #: 77126-514
Supplier: Prosci
New Product
CAS Number:
Specifications
- Gene ID:43740578
- Packaging:Vial
- Protein/peptide type:Recombinant
- Source:Mammalian cells
- Species:SARS-CoV-2
- Conjugation:Unconjugated
- Size:0.1 mg
- Storage conditions:Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2...8 °C for one week . Store at −20...−80 °C for twelve months from the date of receipt.
- Protein synonyms:S glycoprotein RBDSpike glycoprotein Receptor-binding domainDelta variant spike RBDSpike protein RBDS protein RBD
- Protein/peptide name:Spike
- Purity:>90% as determined by SDS-PAGE
- Molecular weight:139.62 kDa
- Sequence:The target gene encoding Ser13-Lys1208. Mutation points A67V, H69del, V70del, T95I, G142D, V143del, Y144del, Y145del, N211del, L212I, ins214EPE, G339D, S371L, S373P, S375F, K417N, N440K, G446S, S477N, T478K, E484A, Q493R, G496S, Q498R, N501Y, Y505H, T547K, D614G, H655Y, N679K, P681H, N764K, D796Y, N856K, Q954H, N969K, L981F
- Formulation:Lyophilized. Supplied as solution form in PBS, pH 7.5 or Lyophilized from PBS, pH 7.5
- Shipping temperature:Room temperature
Specifications
About this item
Customers Who Bought This Also Bought