SARS-CoV-2 (COVID-19) Recombinant Omicron Variant (B.1.1.529) Spike RBD Protein (from Mammalian Cells)
Catalog #: 77129-464
Supplier: Prosci
New Product
CAS Number:
Specifications
- Gene ID:43740578
- Packaging:Vial
- Protein/peptide type:Recombinant
- Source:Mammalian cells
- Species:SARS-CoV-2
- Conjugation:Unconjugated
- Size:0.1 mg
- Storage conditions:Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2...8 °C for one week . Store at −20...−80 °C for twelve months from the date of receipt.
- Protein synonyms:S glycoprotein RBDSpike glycoprotein Receptor-binding domainDelta variant spike RBDSpike protein RBDS protein RBD
- Protein/peptide name:Spike
- Purity:>90% as determined by SDS-PAGE
- Molecular weight:53.21 kDa
- Sequence:RBD with mutations at G339D, S371L, S373P, S375F, K417N, N440K, G446S, S477N, T478K, E484A, Q493K, G496S, Q498R, N501Y, Y505H
- Formulation:Lyophilized. Supplied as solution form in PBS, pH 7.5 or Lyophilized from PBS, pH 7.5
- Shipping temperature:Room temperature
Specifications
About this item
Customers Who Bought This Also Bought