SARS-CoV-2 (COVID-19, 2019-nCoV) Recombinant Alpha Variant (B.1.1.7, UK) Spike RBD Protein (from HEK293 cells)
Catalog #: 77124-026
Supplier: Prosci
New Product
CAS Number:
Specifications
- Gene ID:43740568
- Packaging:Vial
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:SARS-CoV-2
- Conjugation:Unconjugated
- Size:0.1 mg
- Storage conditions:–80 °C. Avoid freeze-thaw cycles.
- Biological activity:Not determined
- Protein synonyms:Spike proteinSurface GlycoproteinSARS-CoV-2 (COVID-19) Spike RBD Recombinnat Protein: Severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2)
- Protein/peptide name:Spike
- Purity:>98% as determined by SDS-PAGE. PureCube Indigo Agarose.
- Molecular weight:225 amino acids (including His tag); 25506 Da
- Sequence:sequence (aa 319 – 537), N501Y C-terminal His tag
- Concentration:Batch dependent
- Formulation:Liquid. 20 mM Hepes pH 7.5; 150 mM NaCl
- Shipping temperature:Dry ice
- Tested applications:ELISA
Specifications
About this item
Customers Who Bought This Also Bought