SARS-CoV-2 (COVID-19, 2019-nCoV) Recombinant Full-Length Gamma Variant (P.1, Brazil) Spike Protein (from HEK293 cells)
Catalog #: 77127-982
Supplier: Prosci
New Product
CAS Number:
Specifications
- Gene ID:43740568
- Packaging:Vial
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:SARS-CoV-2
- Conjugation:Unconjugated
- Size:0.025 mg
- Storage conditions:–80 °C. Avoid freeze-thaw cycles.
- Biological activity:Not determined
- Protein synonyms:SARS-CoV-2 (COVID-19) Spike Recombinant Protein: Severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2)Spike proteinSurface Glycoprotein
- Protein/peptide name:Spike
- Purity:>98% as determined by SDS-PAGE. PureCube Rho1D4 Agarose.
- Molecular weight:1286 amino acids (including Rho1D4 tag and linker); 142114 Da
- Sequence:Full-length sequence (aa 1 – 1273), L18F, T20N, P26S, D138Y, R190S, K417T, E484K, N501Y, H655Y, T1027I, V1176F furin cleavage site “RRAR” mutated to “GSAG”; KV986PP C-terminal Rho1D4 tag fused with spacer “GSSG” to protein sequence
- Concentration:Batch dependent
- Formulation:Liquid. 20 mM Hepes pH 7.5; 150 mM NaCl, 0.001% LMNG
- Shipping temperature:Dry ice
- Tested applications:ELISA
Specifications
About this item
Customers Who Bought This Also Bought