SARS-CoV-2 (COVID-19, 2019-nCoV) Recombinant full length spike protein in DIBMA Glycerol (from HEK293 cells)
Catalog #: 77132-292
Supplier: Prosci
New Product
CAS Number:
Specifications
- Gene ID:43740568
- Packaging:Vial
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:SARS-CoV-2
- Conjugation:Unconjugated
- Size:0.025 mg
- Storage conditions:–80 °C. Avoid freeze-thaw cycles.
- Biological activity:Not determined
- Protein synonyms:SARS-CoV-2 (COVID-19) Spike Recombinant Protein: Severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2)Spike proteinSurface Glycoprotein
- Protein/peptide name:Spike
- Purity:>98% as determined by SDS-PAGE. PureCube Rho1D4 Agarose.
- Molecular weight:1286 amino acids (including Rho1D4 tag and linker); 142114 Da
- Sequence:Full-length sequence (aa 1 – 1273), furin cleavage site “RRAR” mutated to “GSAG”; C-terminal Rho1D4 tag fused with spacer “GSSG” to protein sequence
- Concentration:Batch dependent
- Formulation:Liquid. 20 mM Hepes pH 7.5; 150 mM NaCl,: buffer does not contain free DIBMA Glycerol (DIBMA: Diisobutylene-maleic acid)
- Shipping temperature:Dry ice
- Tested applications:ELISA
Specifications
About this item
Customers Who Bought This Also Bought