Human Recombinant SARS-CoV-2 (COVID-19) Spike (D614G) (Stable Trimer) Protein (from HEK293 cells)
Catalog #: 77133-676
Supplier: Prosci
New Product
CAS Number:
Specifications
- Gene ID:43740568
- Packaging:Vial
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Conjugation:Unconjugated
- Size:50 µg
- Storage conditions:Short term storageL: 4 °C, long term storage: −20 °C, Handling advice: After opening, prepare aliquots and store at −20 °C. Avoid freeze/thaw cycles. Centrifuge lyophilized vial before opening and reconstitution. For maximum product recovery after thawing, centrifuge the vial before opening the cap. Use/Stability: Stable for at least 6 months after receipt when stored at −20 °C. Working aliquots are stable for up to 3 months when stored at −20 °C.
- Biological activity:Binds to the human SARS-CoV-2 receptor ACE2. Binds to anti-SARS-CoV-2 Spike antibodies in serum or plasma.
- Protein synonyms:2019-nCoV Spike Protein S
- Protein/peptide name:Spike
- Purity:≥90% (SDS-PAGE)
- Molecular weight:~190 kDa (SDS-PAGE)
- Sequence:Full-length soluble SARS-CoV-2 Spike protein (aa 16-1213) including a foldon trimerization motif, mutated Furin recognition site (R682S, R685S) and two stabilizing mutations (K986P and V987P) is fused at the C-terminus to a His-tag.
- Concentration:0.2 mg/ml after reconstitution
- Formulation:Lyophilized. Lyophilized. Contains PBS and 0.5% trehalose.
- Shipping temperature:Room temperature
Specifications
About this item
Customers Who Bought This Also Bought