Order Entry
ContactUsLinkComponent
Human Recombinant SARS-CoV-2 (COVID-19) Beta Variant (B.1.351, SA) Spike S1 (RBD) Protein (from HEK293 cells)
Human Recombinant SARS-CoV-2 (COVID-19) Beta Variant (B.1.351, SA) Spike S1 (RBD) Protein (from HEK293 cells)
Catalog #: 77119-416
Supplier:  Prosci
New Product
CAS Number:  
Human Recombinant SARS-CoV-2 (COVID-19) Beta Variant (B.1.351, SA) Spike S1 (RBD) Protein (from HEK293 cells)
Catalog #: 77119-416
Supplier:  Prosci
New Product
CAS Number:  

Specifications

  • Gene ID:
    43740568
  • Packaging:
    Vial
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Conjugation:
    Unconjugated
  • Size:
    50 µg
  • Storage conditions:
    Short term storageL: +4 °C, Long term storage: −20 °C, Handling Advice: After opening, prepare aliquots and store at −20 °C. Avoid freeze/thaw cycles. Centrifuge lyophilized vial before opening and reconstitution. For maximum product recovery after thawing, centrifuge the vial before opening the cap. Use/Stability: Stable for at least 6 months after receipt when stored at −20 °C. Working aliquots are stable for up to 3 months when stored at −20 °C.
  • Biological activity:
    Binds to human ACE2.
  • Protein synonyms:
    Spike Receptor Binding Domain Variant South Africa
    2019-nCoV Spike Protein S1 (RBD)
    501Y.V2
  • Protein/peptide name:
    Spike
  • Purity:
    ≥95% (SDS-PAGE)
  • Molecular weight:
    ~60 kDa (SDS-PAGE)
  • Sequence:
    Receptor-binding domain (RBD) of SARS-CoV-2 Spike protein S1 (aa 319-541) containing the mutations K417N, E484K, N501Y is fused to the N-terminus of the Fc region of human IgG1.
  • Concentration:
    1 mg/ml after reconstitution
  • Formulation:
    Lyophilized. Lyophilized. Contains PBS.
  • Shipping temperature:
    Room temperature

Specifications

About this item

Customers Who Bought This Also Bought